missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OVOL3 Polyclonal Antibody

Product Code. 18025691
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18025691 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18025691 Supplier Invitrogen™ Supplier No. PA5144818
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: CSLEAHLAKV HGQPASYAYR ERREKLHVCE DCGFTSSRPD TYAQH Antigen sequence identity: Mouse 98%; Rat 96%.

OVOL3 is a transcription factor that plays a crucial role in the development and differentiation of various tissues, including the skin, hair, and eyes. It is involved in regulating the expression of genes that control cell proliferation, differentiation, and apoptosis.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen OVOL3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OVOL3
Gene Accession No. O00110
Gene Alias HOVO; OVO3; OVO-like zinc finger 3; Zinc finger protein OVO-like 3
Gene Symbols OVOL3
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 163-207. Recombinant protein control fragment (Product #RP-110085).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728361
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.