missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRS2 Antibody (SR2015), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21759-50ul
This item is not returnable.
View return policy
Description
FRS2 Monoclonal antibody specifically detects FRS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
Specifications
| FRS2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| SR2015 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:20, Immunoprecipitation 1:50 | |
| FGFR signalling adaptor, FGFR-signaling adaptor SNT, fibroblast growth factor receptor substrate 2, FRS2A, FRS2alpha, SNT, SNT1, SNT-1FGFR substrate 2, Suc1-associated neurotrophic factor target 1 | |
| A synthesized peptide derived from human FRS2 (Uniprot #: Q8WU20) | |
| 50 μg | |
| Cancer, Signal Transduction | |
| 10818 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction