missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-83421-25ul
This item is not returnable.
View return policy
Description
FRS3 Polyclonal specifically detects FRS3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FRS3 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| FGFR substrate 3, FGFR-signaling adaptor SNT2, fibroblast growth factor receptor substrate 3, FRS2B, FRS2beta, FRS2-beta, SNT2, SNT-2MGC17167, Suc1-associated neurotrophic factor target 2, suc1-associated neurotrophic factor target 2 (FGFR signalling adaptor) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FRS3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EEPVIITRNSHPAELDLPRAPQPPNALGYTVSSFSNGCPGEGPRFSAPRRLSTSSLRHPSLGEESTHALIAPDEQSHTYVNTPASEDDHR | |
| 25 μL | |
| Signal Transduction | |
| 10817 | |
| Human | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur