missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FSCN2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
180.00 € - 453.00 €
Specifications
| Antigen | FSCN2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17743374
|
Invitrogen™
PA5121976 |
20 μL |
180.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17733374
![]() |
Invitrogen™
PA5121975 |
100 μL |
453.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FSCN2 acts as an actin bundling protein. May play a pivotal role in photoreceptor cell-specific events, such as disk morphogenesis.Specifications
| FSCN2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O14926 | |
| 25794 | |
| Gln8-His258 (Accession O14926), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| FSCN2 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| A930022G03; Ahl8; C630046B20Rik; fascin 2; fascin 2, retinal; fascin actin-bundling protein 2, retinal; fascin homolog 2, actin-bundling protein, retinal; fascin homolog 2, actin-bundling protein, retinal (Strongylocentrotus purpuratus); fascin-2; Fscn2; Retinal fascin; RFSN; RP30 | |
| FSCN2 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title