Learn More
FSTL1 Mouse, anti-Human, Clone: 1G5G1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68528-1-IG-150UL
 View more versions of this product
Product Code. 18024785
  • 410.00 € / 150µL
Description
The FSTL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FSTL1. This antibody recognizes human, mouse, rat antigen. The FSTL1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
FSTL1
1G5G1
FLJ50214, FLJ52277, follistatin like 1, Follistatin like protein 1, Follistatin related protein 1, FRP, FSL1, FSTL1
150 μL
11167
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat
150 kDa
IgG2a