Learn More
FSTL1 Mouse, anti-Human, Clone: 1G5G1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68528-1-IG-20UL
This item is not returnable.
View return policy
Description
The FSTL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FSTL1. This antibody recognizes human, mouse, rat antigen. The FSTL1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| FSTL1 | |
| 1G5G1 | |
| FLJ50214, FLJ52277, follistatin like 1, Follistatin like protein 1, Follistatin related protein 1, FRP, FSL1, FSTL1 | |
| 20 μL | |
| 11167 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG2a |