missing translation for 'onlineSavingsMsg'
Learn More

FT011, MedChemExpress

Product Code. 16474346
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474346 100 mg 100mg
16490016 2 mg 2mg
16443048 10 mg 10mg
16415787 5 mg 5mg
16409616 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16474346 Supplier Medchem Express Supplier No. HY100495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FT011 is an anti-fibrotic agent, reduces mRNA expression of collagens I and III and inhibits collagen synthesis.

FT011 is an anti-fibrotic agent, reduces mRNA expression of collagens I and III and inhibits collagen synthesis.

Specifications

Chemical Name or Material FT011
CAS 1001288-58-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H17NO5
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (284.62 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=CC=C1NC(/C=C/C2=CC=C(OCC#C)C(OC)=C2)=O
Molecular Weight (g/mol) 351.35
Formula Weight 351.35
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.