missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUCA2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00 € - 349.00 €
Specifications
| Antigen | FUCA2 |
|---|---|
| Concentration | 400 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235705
|
Proteintech
30080-1-AP-20UL |
20 μL |
133.74 € 129.00 € / 20µL Save 4.74 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235213
|
Proteintech
30080-1-AP-150UL |
150 μL |
361.81 € 349.00 € / 150µL Save 12.81 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FUCA2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FUCA2. This antibody recognizes human antigen. The FUCA2 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| FUCA2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| MGC1314; dJ20N2.5 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 400 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 2519 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title