missing translation for 'onlineSavingsMsg'
Learn More

Fucose Mutarotase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

216.00 € - 326.00 €

Specifications

Antigen Fucose Mutarotase
Dilution ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18613788
View Documents
Novus Biologicals
NBP3-00239-100ul
100 μg
326.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18682399
View Documents
Novus Biologicals
NBP3-00239-50ul
50 μg
216.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fucose Mutarotase Polyclonal antibody specifically detects Fucose Mutarotase in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

Fucose Mutarotase
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
C10orf125, chromosome 10 open reading frame 125, FucM, fucose mutarotase, FUCU
Produced in rabbits immunized with purified, recombinant Human Fucose Mutarotase (Accession#: A2VDF0-1; Met1-Leu154)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000
Polyclonal
Purified
RUO
0.2 um filtered solution in PBS
282969
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.