missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fukutin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18276-1-AP-20UL
This item is not returnable.
View return policy
Description
FKTN regulates the migration and assembly of neurons during cortical histogenesis. Fukuyama congenital muscular dystrophy results from mutations in its gene.Specifications
| Fukutin | |
| Polyclonal | |
| Unconjugated | |
| FKTN | |
| CMD1X, FCMD, FKTN, Fukutin, LGMD2M | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2218, 246179, 362520 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75072, Q8R507 | |
| Fktn | |
| Fukutin Fusion Protein Ag13067 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction