missing translation for 'onlineSavingsMsg'
Learn More

EKASTU Safety Full Face Mask

Product Code. 10743923
Change view
Click to view available options
Description:
C 607/F (Class 3); 2 Exhalation valves
C 607/Silicone (Class 2); 1 Exhalation valve
SFERA (Class 3); 1 Exhalation valve
SFERA/Silicone (Class 3); 2 Exhalation valves
Color:
Black
Yellow
Material (Faceshield):
EPDM
Silicone
Material (Lens):
Polycarbonate
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Color Material (Faceshield) Material (Lens) Unit Size
10743923 SFERA/Silicone (Class 3); 2 Exhalation valves Yellow Silicone Polycarbonate -
10077850 C 607/F (Class 3); 2 Exhalation valves Black EPDM Polycarbonate Each
10117030 SFERA (Class 3); 1 Exhalation valve Black EPDM Polycarbonate Each
10581412 C 607/Silicone (Class 2); 1 Exhalation valve Yellow Silicone Polycarbonate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10743923 Supplier EKASTU Safety Supplier No. 466618
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

EKASTU Full Face Mask with anti-fogging, non-reflecting, distortion-free polycarbonate visor with large field of vision.

Features:

  • Built-in speech diaphragm
  • Outstanding transmission of voice allows the use of megaphones, telephones and two-way radios
  • Uuniversal size allows the mask to fit tightly, but without pressure, irrespective of the face shape
  • Exhalation valves with exhalation valve pre-chamber results in low exhalation resistance and prevents dangerous media from penetrating the inside of the mask
  • Inner mask is equipped with 2 check valves to reduce clearance volume and prevent the visor shield from misting up

Specifications

Certifications/Compliance DIN EN 136
Connection Type Standard thread DIN EN 148-1
Description SFERA/Silicone (Class 3); 2 Exhalation valves
Color Yellow
Disposable Reusable
Product Type Full Face Mask
Size One Size
Facepiece Type Anti-fogging, non-reflecting, distortion-free
For Use With (Equipment) In connection with respiratory filter (standard thread DIN EN 148-1)
Lens Type Visor
Material (Faceshield) Silicone
Material (Lens) Polycarbonate
Resistant To Anti-scratch
Weight (Metric) Approx. 620 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.