missing translation for 'onlineSavingsMsg'
Learn More

Fullerene C60, TRC

Product Code. 30134395
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134395 250 mg 250mg
30133224 100 mg 100mg
17933355 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134395 Supplier Toronto Research Chemicals Supplier No. TRCF862543250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99685-96-8
Molecular Formula C60
Molecular Weight (g/mol) 720.64
Synonym Buckminsterfullerene, Buckminsterfullerene (C60), Buckyball, C60 Fullerene, Carbon (C60), Carbon (C60) fullerene, Carbon (C60) mol., Carbon cluster (C60), Carbon, mol. (C60), Follene-60, Footballene, Footballene (C60), Fullerene, Fullerene C60 cluster, Fullerene-60, Fullerene-C60, Icosahedral C60, Nano-C60, Nanom Purple, Nanom Purple N 60S, Nanom Purple ST, Nanom Purple ST-A, Nanom Purple SU, Soccerballene, Tris-PC61BOE, XFC 01, [5,6]Fullerene C60, [60]Fullerene
SMILES c12c3c4c5c6c7c8c9c%10c%11c%12c9c%13c%14c%15c%12c%16c%17c%11c%18c%19c%10c%20c8c6c%21c%22c%20c%19c%23c%24c%18c%17c%25c%26c%16c%15c%27c%28c%14c(c7%13)c5c3c%28c%29c%27c%26c%30c%25c%24c%31c%23c%22c(c4%21)c1c%31c%30c2%29

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C60/c1-2-5-6-3(1)8-12-10-4(1)9-11-7(2)17-21-13(5)23-24-14(6)22-18(8)28-20(12)30-26-16(10)15(9)25-29-19(11)27(17)37-41-31(21)33(23)43-44-34(24)32(22)42-38(28)48-40(30)46-36(26)35(25)45-39(29)47(37)55-49(41)51(43)57-52(44)50(42)56(48)59-54(46)53(45)58(55)60(57)59
Formula Weight 720
Chemical Name or Material Fullerene C60
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.