missing translation for 'onlineSavingsMsg'
Learn More

Fullerene C60, TRC

Product Code. 17933355
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933355 50 mg 50mg
30133224 100 mg 100mg
30134395 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933355 Supplier Toronto Research Chemicals Supplier No. TRCF86254350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99685-96-8
Formula Weight 720
InChI Formula InChI=1S/C60/c1-2-5-6-3(1)8-12-10-4(1)9-11-7(2)17-21-13(5)23-24-14(6)22-18(8)28-20(12)30-26-16(10)15(9)25-29-19(11)27(17)37-41-31(21)33(23)43-44-34(24)32(22)42-38(28)48-40(30)46-36(26)35(25)45-39(29)47(37)55-49(41)51(43)57-52(44)50(42)56(48)59-54(46)53(45)58(55)60(57)59
Molecular Formula C60
Molecular Weight (g/mol) 720.64
SMILES c12c3c4c5c6c7c8c9c%10c%11c%12c9c%13c%14c%15c%12c%16c%17c%11c%18c%19c%10c%20c8c6c%21c%22c%20c%19c%23c%24c%18c%17c%25c%26c%16c%15c%27c%28c%14c(c7%13)c5c3c%28c%29c%27c%26c%30c%25c%24c%31c%23c%22c(c4%21)c1c%31c%30c2%29
Synonym Buckminsterfullerene, Buckminsterfullerene (C60), Buckyball, C60 Fullerene, Carbon (C60), Carbon (C60) fullerene, Carbon (C60) mol., Carbon cluster (C60), Carbon, mol. (C60), Follene-60, Footballene, Footballene (C60), Fullerene, Fullerene C60 cluster, Fullerene-60, Fullerene-C60, Icosahedral C60, Nano-C60, Nanom Purple, Nanom Purple N 60S, Nanom Purple ST, Nanom Purple ST-A, Nanom Purple SU, Soccerballene, Tris-PC61BOE, XFC 01, [5,6]Fullerene C60, [60]Fullerene
Chemical Name or Material Fullerene C60

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.