missing translation for 'onlineSavingsMsg'
Learn More

Fullerene powder, 99.9+% C{60}

Product Code. 11365027
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11365027 250 mg 250mg
11345027 1 g 1g
11355027 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11365027 Supplier Thermo Scientific Alfa Aesar Supplier No. 042007.MD
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99685-96-8 | C60 | 720.66 g/mol

Fullerene powder is useful for biomedical applications such as the design of high-performance MRI contrast agents, x-ray imaging contrast agents and photodynamic therapy. It is also essential for single-walled carbon nanotubes with narrow diameter-distribution. It is used as light-activated antimicrobial agent. It also finds application in the field of nanotechnology, heat resistance and superconductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 99685-96-8
Molecular Formula C60
Molecular Weight (g/mol) 720.66
MDL Number MFCD00282904
InChI Key XMWRBQBLMFGWIX-UHFFFAOYSA-N
Synonym fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
PubChem CID 123591
ChEBI CHEBI:33128
IUPAC Name (C\{60}-I\{h})[5,6]fullerene
SMILES C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41

Tekniske data

Density 1.7 g/mL
Boiling Point 500°C to 600°C (sublimation)
Assay Percent Range >99.9% C60
Quantity 250 mg
UN Number UN1325
Solubility Information Soluble in toluene. Insoluble in water.
Percent Purity ≥99.9%
Physical Form Powder
Assay C60
Chemical Name or Material Fullerene powder

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.