missing translation for 'onlineSavingsMsg'
Learn More

Fullerene powder, mixed, typically 98% C{60}, 2% C{70}

Product Code. 11365297
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11365297 250 mg 250mg
11355297 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365297 Supplier Thermo Scientific Alfa Aesar Supplier No. 040970.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99685-96-8 | C60 | 720.66 g/mol

The optical absorption properties of C60 match solar spectrum in a way that suggests that C60-based films could be useful for photovoltaic applications. Because of its high electronic affinity it is one of the most common electron acceptor used in donor/acceptor based solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99685-96-8
Molecular Formula C60
Molecular Weight (g/mol) 720.66
MDL Number MFCD00282904
InChI Key XMWRBQBLMFGWIX-UHFFFAOYSA-N
Synonym fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
PubChem CID 123591
ChEBI CHEBI:33128
IUPAC Name (C\{60}-I\{h})[5,6]fullerene
SMILES C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41

Specifications

Color Black
Density ≈ 1.7 g/mL
Boiling Point 500°C to 600°C (sublimation)
Quantity 250 mg
UN Number UN1325
Solubility Information Soluble in organic solvents. Insoluble in water.
Concentration or Composition (by Analyte or Components) Typically 98% C60, 2% C70
Physical Form Powder
Chemical Name or Material Fullerene powder, Mixed

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.