missing translation for 'onlineSavingsMsg'
Learn More

Fulvestrant, MedChemExpress

Product Code. 16401947
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401947 50 mg 50mg
16455788 10 mg 10mg
16406266 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16401947 Supplier Medchem Express Supplier No. HY13636
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fulvestrant (ICI 182780) is a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant is also a GPR30 agonist. Fulvestrant effectively inhibits the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM. Fulvestrant also induces autophagy and has antitumor efficacy.

Fulvestrant (ICI 182780) is a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant is also a GPR30 agonist. Fulvestrant effectively inhibits the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM. Fulvestrant also induces autophagy and has antitumor efficacy.

Specifications

Chemical Name or Material Fulvestrant
CAS 129453-61-8
Color White
Purity Grade Notes Research
Molecular Formula C32H47F5O3S
Quantity 50 mg
Synonym ICI 182780 ZD 9238 ZM 182780
Solubility Information DMSO : 250 mg/mL (412.02 mM; Need ultrasonic)
SMILES C[C@@]12[C@@H](O)CC[C@@]1([H])[C@]3([H])[C@H](CCCCCCCCCS(CCCC(F)(F)C(F)(F)F)=O)CC4=C(C=CC(O)=C4)[C@@]3([H])CC2
Molecular Weight (g/mol) 606.77
Formula Weight 606.77
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.