missing translation for 'onlineSavingsMsg'
Learn More

Fumaric acid-dimethyl ester 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30316994
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316994 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316994 Supplier Dr. Ehrenstorfer Supplier No. DREA13955800AL1000
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fumaric acid dimethyl ester
CAS 624-49-7
Analyte or Component Names Fumaric acid dimethyl ester
Shipping Condition Room Temperature
Molecular Formula C6 H8 O4
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym (2E)-2-Butenedioic Acid 1,4-Dimethyl Ester, (E)-2-Butenedioic Acid 1,4-Dimethyl Ester, trans-1,2-Ethylenedicarboxylic Acid, Fumaric Acid Dimethyl Ester, (2E)-2-Butenedioic Acid Dimethyl Ester, (E)-But-2-enedioic Acid Dimethyl Ester, AZL-O 211089, Allomaleic Acid Dimethyl Ester, BG 00012, Boletic Acid Dimethyl Ester, Dimethyl (E)-2-Butenedioate, Dimethyl (E)-Butenedioate, Dimethyl Fumarate, Dimethyl trans-Ethylenedicarboxylate, Methyl Fumarate, NSC 167432, NSC 25942, trans-1,2-Ethylenedicarboxylic Acid Dimethyl Ester, trans-Butenedioic Acid Dimethyl Ester, Fumaric acid dimethyl ester
InChI Formula InChI=1S/C6H8O4/c1-9-5(7)3-4-6(8)10-2/h3-4H,1-2H3/b4-3+
SMILES COC(=O)\C=C\C(=O)OC
IUPAC Name dimethyl (E)-but-2-enedioate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 144.13
Quantity 1 mL
Formula Weight 144.0423 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.