missing translation for 'onlineSavingsMsg'
Learn More

Fumaric acid, Dr. Ehrenstorfer

Product Code. 30314450
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314450 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314450 Supplier Dr. Ehrenstorfer Supplier No. DREC13955500100MG
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fumaric acid
CAS 110-17-8
Analyte or Component Names Fumaric acid
Shipping Condition +20°C
Molecular Formula C4 H4 O4
Certifications/Compliance ISO 17034
Synonym (E)-Butenedioic Acid, Fumaric Acid, Malic Acid Imp. A (EP), Aspartic Acid Imp. B (Pharmeuropa), Aspartic Acid Imp. B (EP), Imp. A (EP), (2E)-But-2-enedioic Acid, Asparagine Monohydrate Impurity D, Aspartic Acid Impurity B, Magnesium Aspartate Dihydrate Impurity C, Malic Acid Impurity A, Asparagine Impurity D, Magnesium Aspartate Impurity C
InChI Formula InChI=1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+
SMILES OC(=O)\C=C\C(=O)O
IUPAC Name (E)-but-2-enedioic acid
Molecular Weight (g/mol) 116.07
Quantity 100 mg
Formula Weight 116.011 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.