missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FYCO1 Monoclonal Antibody (OTI9D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809806
This item is not returnable.
View return policy
Description
This gene encodes a protein that contains a RUN domain, FYVE-type zinc finger domain and Golgi dynamics (GOLD) domain. The encoded protein plays a role in microtubule plus end-directed transport of autophagic vesicles through interactions with the small GTPase Rab7, phosphatidylinositol-3-phosphate (PI3P) and the autophagosome marker LC3. Mutations in this gene are a cause of autosomal recessive congenital cataract-2 (CATC2).Specifications
| FYCO1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQS8 | |
| FYCO1 | |
| Human recombit protein fragment corresponding to amino acids 1102-1187 of human FYCO1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI9D1 | |
| Unconjugated | |
| FYCO1 | |
| CATC2, CTRCT18, RUFY3, ZFYVE7 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79443 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction