missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FYCO1 Monoclonal Antibody (OTI9D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809806S
This item is not returnable.
View return policy
Description
FYCO1 Monoclonal antibody specifically detects FYCO1 in Human samples. It is validated for Western BlotSpecifications
| FYCO1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17980 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5702 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI9D1 | |
| Unconjugated | |
| PSMC3 | |
| 2810409M01Rik; CATC2; CTRCT18; DKFZp779K1152; FLJ13335; FYCO1; FYVE and coiled-coil domain autophagy adaptor 1; FYVE and coiled-coil domain containing 1; FYVE and coiled-coil domain-containing protein 1; maternal embryonic message 2; Mem2; MGC126517; MGC126519; RUFY3; RUN and FYVE domain containing 3; ZFYVE7; Zinc finger FYVE domain-containing protein 7 | |
| Human recombinant protein fragment corresponding to amino acids 353-438 of human PSMC3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction