missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fyn Antibody (S07-9I2), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00 € - 645.00 €
Specifications
| Antigen | Fyn |
|---|---|
| Clone | S07-9I2 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618373
|
Novus Biologicals
NBP3-19641-50ul |
50 μg |
421.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627043
|
Novus Biologicals
NBP3-19641-100ul |
100 μg |
645.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fyn Monoclonal antibody specifically detects Fyn in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Fyn | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.10, EC 2.7.10.2, FYN oncogene related to SRC, FGR, YES, MGC45350, OKT3-induced calcium influx regulator, p59-Fyn, protein-tyrosine kinase fyn, Proto-oncogene c-Fyn, Proto-oncogene Syn, proto-oncogene tyrosine-protein kinase fyn, SLKc-syn protooncogene, src/yes-related novel, Src-like kinase, SYN, tyrosine kinase p59fyn(T), tyrosine-protein kinase Fyn | |
| A synthetic peptide of mouse Fyn (Uniprot # P39688) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S07-9I2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 2534 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title