missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FZD4 Recombinant Rabbit Monoclonal Antibody (HL2824)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556717
This item is not returnable.
View return policy
Description
FZD4 Recombinant Monoclonal Antibody for Western Blot
FZD4 is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. This protein may play a role as a positive regulator of the Wingless type MMTV integration site signaling pathway. A transcript variant retaining intronic sequence and encoding a shorter isoform has been described, however, its expression is not supported by other experimental evidence. Frizzled 4 is widely expressed, particularly in heart, skeletal muscle, ovary, and fetal kidney. Transcripts have also been identified in liver, kidney, pancreas, spleen, and fetal lung, and in smaller amounts in placenta, adult lung, prostate, testis, colon, and fetal brain.
Specifications
| FZD4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9ULV1 | |
| FZD4 | |
| Synthetic peptide encompassing a sequence within the Intracellular domain of human Frizzled 4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Zebrafish | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2824 | |
| Unconjugated | |
| FZD4 | |
| CD344; CD344 antigen; EVR1; FEVR; frizzled 4, seven transmembrane spanning receptor; frizzled class receptor 4; frizzled family receptor 4; frizzled homolog 4; frizzled homolog 4 (Drosophila); frizzled receptor 4; frizzled-4; Fz4; Fz-4; Fzd4; FZD4S; FzE4; GPCR; hFz4; mFz4; MGC34390; rFz4; WNT receptor frizzled-4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 30363, 8322 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction