missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-CSFR/CD114 Antibody (723806), PE/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB60391PECY7
This item is not returnable.
View return policy
Description
G-CSFR/CD114 Monoclonal antibody specifically detects G-CSFR/CD114 in Mouse samples. It is validated for Flow Cytometry
Specifications
| G-CSFR/CD114 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1441 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry | |
| 723806 | |
| Flow Cytometry | |
| CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), G-CSF receptor, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor | |
| Mouse myeloma cell line NS0-derived recombinant mouse G-CSF R/CD114, Cys26-Asp626, Accession # P40223 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction