missing translation for 'onlineSavingsMsg'
Learn More

G3BP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

378.00 € - 480.00 €

Specifications

Antigen G3BP1
Dilution Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18439800
View Documents
Novus Biologicals
NBP1-83404-25ul
25 μL
378.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18791324
View Documents
Novus Biologicals
NBP1-83404
0.1 mL
480.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

G3BP1 Polyclonal specifically detects G3BP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

G3BP1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat
ATP-dependent DNA helicase VIII, EC 3.6.1, EC 3.6.4.12, EC 3.6.4.13, G3BP-1, G3BPRas-GTPase-activating protein SH3-domain-binding protein, GAP binding protein, GAP SH3 domain-binding protein 1, GTPase activating protein (SH3 domain) binding protein 1, hDH VIII, HDH-VIII, MGC111040, ras GTPase-activating protein-binding protein 1, RasGAP-associated endoribonuclease G3BP
G3BP1
IgG
Affinity Purified
Specificity of human G3BP1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated
Polyclonal
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
10146
This antibody was developed against Recombinant Protein corresponding to amino acids:FRYQDEVFGGFVTEPQEESEEEVEEPEERQQTPEVVPDDSGTFYDQAVVSNDMEEHLEEPVAEPEPDPEPEPEQEPVSEIQEEKPEPVLEETAP
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.