Learn More
Invitrogen™ G3BP1 Recombinant Rabbit Monoclonal Antibody (24GB4575)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557404
 View more versions of this product
Product Code. 30276449
  • 564.00 € / 100µL
Description
G3BP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes one of the DNA-unwinding enzymes which prefers partially unwound 3'-tailed substrates and can also unwind partial RNA/DNA and RNA/RNA duplexes in an ATP-dependent fashion. This enzyme is a member of the heterogeneous nuclear RNA-binding proteins and is also an element of the Ras signal transduction pathway. It binds specifically to the Ras-GTPase-activating protein by associating with its SH3 domain. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
G3BP1
Recombinant Monoclonal
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q13283
G3BP1
A synthesized peptide derived from human G3BP (aa 219-231).
100 μL
Primary
Human, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4575
Unconjugated
G3BP1
AI849976; ATP-dependent DNA helicase VIII; B430204O07; C87777; DNA helicase VIII; G3BP; G3BP stress granule assembly factor 1; G3bp1; G3BP-1; GAP binding protein; GAP SH3 binding protein; GAP SH3 domain-binding protein 1; GTPase activating protein (SH3 domain) binding protein 1; hDH VIII; HDH-VIII; MGC111040; mKIAA4115; Ras GTPase-activating protein-binding protein 1; RasGAP-associated endoribonuclease G3BP; Ras-GTPase-activating protein SH3-domain binding protein 1; Ras-GTPase-activating protein SH3-domain-binding protein; RGD1310666
Rabbit
Affinity chromatography
RUO
10146, 171092
-20°C