missing translation for 'onlineSavingsMsg'
Learn More

G3BP2 Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech

Product Code. 17751349 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17751349 20 μL 20µL
17689896 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17751349 Supplier Proteintech Supplier No. CL5551627620UL
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

G3BP2 Polyclonal antibody apecifically detetects G3BP2 antigen in Human Samples. It is validated for Immunofluorescence.

Specifications

Antigen G3BP2
Applications Immunofluorescence
Classification Polyclonal
Concentration 1000 ug/mL
Conjugate CoraLite 555
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC011731
Gene Symbols G3BP2
Host Species Rabbit
Immunogen G3BP2 fusion protein Ag9355
Purification Method Antigen affinity purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 9908
Target Species Human
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.