missing translation for 'onlineSavingsMsg'
Learn More

G418 disulfate, Cell Culture Reagent

Product Code. 15434999
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434999 1 g 1g
15444999 5 g 5g
15454999 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15434999 Supplier Thermo Scientific Alfa Aesar Supplier No. J62671.03
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

G418 sulfate, geneticin, CAS # 108321-42-2, is an antibiotic closely related to Gentamicin B, the common aminoglycoside antibiotic. | CAS: 108321-42-2 | C20H44N4O18S2 | 692.70 g/mol

G418 disulfate is an aminoglycoside that is toxic to bacteria, yeast, protozoans and helminths. It prevents protein synthesis by blocking the elongation step in prokaryotic and eukaryotic ribosomes. It is used as a selection agent for G418 resistant bacteria or fungi after transformation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• G418 sulfate (geneticin) is an aminoglycoside antibiotic isolated from Micromonospora rhodorangea
• G418 is a bacteriostatic that prevents polypeptide formation by inhibiting the elongation step of polypeptide synthesis
Applications
• G418 sulfate can be used for selection of genetically engineered cells in both eukaryotes and prokaryotes
• Laboratory grade for applications such as biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108321-42-2
Molecular Formula C20H44N4O18S2
Molecular Weight (g/mol) 692.70
MDL Number MFCD00058314
InChI Key UHEPSJJJMTWUCP-KETIWVBHNA-N
Synonym Geneticin disulfate
PubChem CID 134129582
SMILES OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O

Specifications

Type G418 Disulfate
Physical Form Powder
Color White
Quantity 1 g
Solubility Information Soluble in water.
IUPAC Name (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
Formula Weight 692.71
Grade Cell Culture
Chemical Name or Material G418 disulfate
compliance-icons
Product Identifier
  • G418 disulfate, Cell Culture Reagent
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: D-Streptamine, O-2-amino-2,7-dideoxy-D-glycero-.alpha.-D -gluco-heptopyranosyl-(1->4)-O-[3-deoxy-4-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.