missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ G418 Disulfate PhytoPure™

Product Code. 17887496 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17887496 5 g 5g
17670877 250 mg 250mg
11449951 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17887496 Supplier MP Biomedicals™ Supplier No. 0215878294
  • ONLINE EXCLUSIVE PRICE
    853.00 €
    808.50 € / 5g
    Save 44.50 €
    5% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

G418 Disulfate PhytoPure™ is an aminoglycoside related to gentamicin that acts as an antibiotic and is toxic to bacteria, yeast, higher plants, mammalian cells, protozoans and helminths.

  • G418 Disulfate blocks polypeptide synthesis in both prokaryotic and eukaryotic cells and is useful for selection of genetically engineered eukarytoic cells in culture.
  • It is an effective selecting agent utilized in killing prokaryotic and eukaryotic cells.
  • It is also used for selection and maintenance of prokaryotic and eukaryotic cells transfected with a iNOS promoter construct and neomycin resistance gene.

Specifications

CAS 108321-42-2
Color White
Content And Storage The powder should be stored away from light and moisture, store at 2 to 8°C. Solutions should be aliquoted and stored frozen (− 5 to − 20°C).
Form Powder
Product Type Antibiotic
Sterility Non-sterile
Purity or Quality Grade Cell Culture Grade
Quantity 5 g
compliance-icons compliance-icons
Product Identifier
  • Geneticin Disulphate Salt, cell culture grade
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Geneticin DisulphateSalt

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.