missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals G418 sulfate, for biochemistry

Product Code. 10165633 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10165633 5 g Glass Bottle 5g
10143871 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10165633 Supplier Thermo Scientific Chemicals Supplier No. 329400050
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

G418 sulfate, geneticin, CAS # 108321-42-2, is an antibiotic closely related to Gentamicin B, the common aminoglycoside antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• G418 sulfate (geneticin) is an aminoglycoside antibiotic isolated from Micromonospora rhodorangea
• G418 is a bacteriostatic that prevents polypeptide formation by inhibiting the elongation step of polypeptide synthesis


Applications

• G418 sulfate can be used for selection of genetically engineered cells in both eukaryotes and prokaryotes
• Laboratory grade for applications such as biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108321-42-2
Molecular Formula C20H44N4O18S2
Molecular Weight (g/mol) 692.70
MDL Number MFCD00058314
InChI Key UHEPSJJJMTWUCP-KETIWVBHNA-N
PubChem CID 134129582
SMILES OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O

Specifications

Type G418 Sulfate
Physical Form Powder
Infrared Spectrum Authentic
Packaging Glass Bottle
Water 10% max.
Color White
Quantity 5 g
Solubility Information Solubility in water: soluble.
IUPAC Name (2R,3R,4R,5R)-2-[(1S,2R,3R,4S,6R)-4,6-diamino-3-[(2S,3S,4R,5S,6R)-3-amino-4,5-dihydroxy-6-[(1R)-1-hydroxyethyl]oxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
Formula Weight 692.7
Grade Biochemical
Chemical Name or Material G418 sulfate
Show More Show Less
compliance-icons
Product Identifier
  • G418 sulfate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.