missing translation for 'onlineSavingsMsg'
Learn More

G418 Sulfate Solution, MP Biomedicals

Product Code. 11440435 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
20 mL
50 mL
Unit Size:
20mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440435 50 mL 50mL
11430435 20 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440435 Supplier MP Biomedicals Supplier No. 091672548
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

50mg/ml active antibiotic sterile-aqueous solution. Purity: >>98%

Specifications

Type G418 Sulfate
Physical Form Liquid
CAS 108321-42-2, 7732-18-5
Quantity 50 mL
Grade Not Available
Chemical Name or Material G418 Sulfate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.