missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R beta 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
307.00 €
Specifications
| Antigen | GABA-A R beta 2 |
|---|---|
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
GABA-A R beta 2 Polyclonal specifically detects GABA-A R beta 2 in Mouse, Rat samples. It is validated for Western Blot.Specifications
| GABA-A R beta 2 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse | |
| P63138 | |
| 2561 | |
| Fusion protein from the cytoplasmic loop of the beta 2 subunit. Accession # P63138 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 55 kDa |
| Western Blot 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| GABA(A) receptor subunit beta-2, gamma-aminobutyric acid (GABA) A receptor, beta 2, gamma-aminobutyric acid A receptor beta 2, gamma-aminobutyric acid receptor subunit beta-2, MGC119386, MGC119388, MGC119389 | |
| GABRB2 | |
| IgG | |
| Antigen Affinity-purified | |
| Specific for the ∼55k beta 2-subunit of the GABAA receptor in Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title