missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 4 Antibody (S398A-34), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59326
This item is not returnable.
View return policy
Description
GABA-AR alpha 4 Monoclonal specifically detects GABA-AR alpha 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GABA-AR alpha 4 | |
| Monoclonal | |
| Unconjugated | |
| PBS pH7.4, 50% glycerol with 0.1% Sodium Azide | |
| GABRA4 | |
| Fusion protein amino acids 410-450 (Cytoplasmic C-terminus) of rat GABA-A-Receptor Alpha 4 | |
| 100 μg | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2557 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S398A-34 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| GABA(A) receptor subunit alpha-4, gamma-aminobutyric acid (GABA) A receptor, alpha 4, gamma-aminobutyric acid receptor subunit alpha-4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects 60kDa. Does not cross-react with other GABA-A-Receptor Alpha subunits. | |
| Store at -20C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction