missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABARAPL2 Monoclonal Antibody (853746)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA561983
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Involved in intra-Golgi traffic. Modulates intra-Golgi transport through coupling between NSF activity and SNAREs activation. It first stimulates the ATPase activity of NSF which in turn stimulates the association with GOSR1.
Specifications
| GABARAPL2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P60520, P60521, P60522 | |
| GABARAPL2 | |
| E.coli-derived recombinant human GATE16. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 853746 | |
| Unconjugated | |
| GABARAPL2 | |
| 0610012F20Rik; 2900019O08Rik; AI173605; ATG8; ATG8C; FLC3A; GABA type A receptor associated protein like 2; GABA(A) receptor-associated protein like 2; GABA(A) receptor-associated protein-like 2; gabarapl2; gamma-aminobutyric acid (GABA) A receptor-associated protein-like 2; gamma-aminobutyric acid (GABA-A) receptor-associated protein-like 2; gamma-aminobutyric acid receptor-associated protein-like 2; Ganglioside expression factor 2; GATE16; GATE-16; Gef2; GEF-2; Genera; general protein transport factor p16; Golgi-associated ATPase enhancer of 16 kDa; MAP1 light chain 3 related protein; MAP1 light chain 3-related protein; zgc:92319 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 11345, 64670, 93739 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction