missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABRQ Polyclonal Antibody

Description
Peptide sequence: KFHFEFSSAV PEVVLNLFNC KNCANEAVVQ KILDRVLSRY DVRLRPNFGG Sequence homology: Cow: 87%; Dog: 87%; Guinea Pig: 80%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%.
The gamma-aminobutyric acid (GABA) A receptor is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes GABA A receptor, theta subunit. GABRQ gene is mapped to chromosome Xq28 in a cluster including the genes encoding the alpha 3 and epsilon subunits of the same receptor. This gene location is also the candidate region of 2 different neurologic diseases:early-onset parkinsonism (Waisman syndrome) and X-linked mental retardation (MRX3).The gamma-aminobutyric acid (GABA) A receptor is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes GABA A receptor, theta subunit. It is mapped to chromosome Xq28 in a cluster including the genes encoding the alpha 3 and epsilon subunits of the same receptor. This gene location is also the candidate region of 2 different neurologic diseases: early-onset parkinsonism (Waisman syndrome) and X-linked mental retardation (MRX3).
Specifications
Specifications
| Antigen | GABRQ |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | GABRQ |
| Gene Accession No. | Q9UN88 |
| Gene Alias | GABA(A) receptor subunit theta; GABA(A) Theta; GABRQ; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, theta; gamma-aminobutyric acid (GABA) A receptor, subunit theta; gamma-aminobutyric acid (GABA) A receptor, theta; gamma-aminobutyric acid (GABA) receptor, theta; gamma-aminobutyric acid (GABA-A) receptor, subunit theta; gamma-aminobutyric acid A receptor, theta; gamma-aminobutyric acid receptor subunit theta; gamma-aminobutyric acid type A receptor subunit theta; gamma-aminobutyric acid type A receptor theta subunit; THETA |
| Gene Symbols | GABRQ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?