Learn More
GAD1/GAD67 Antibody (CL2914) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46638JF646
Product Code. 30290183
  • 737.00 € / 0.10mL
Description
GAD1/GAD67 Monoclonal antibody specifically detects GAD1/GAD67 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GAD1/GAD67
Monoclonal
Janelia Fluor 646
67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform
This antibody was developed using a recombinant protein derived from Q99259, with the exact immunogen sequence remaining proprietary.
0.1 mL
Diabetes Research
2571
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL2914
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified