missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Antibody (9E10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 33 publications
Brand: Novus Biologicals NB600-302
This item is not returnable.
View return policy
Description
c-Myc Monoclonal specifically detects c-Myc in Human, Mouse, Bovine, Drosophila samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay, Sandwich ELISA, Flow (Intracellular).
Specifications
| c-Myc | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5-2.0 ug/ml, Simple Western 1:200, Flow Cytometry 1:50-1:200. Use reported in scientific literature (PMID 21315712), ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200, Immunoblotting reported in scientific literature (PMID 23750001), Proximity Ligation Assay reported in scientific literature (PMID 33298911), Sandwich ELISA reported in scientific literature (PMID 33122198), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) | |
| 9E10 anti-Myc, 9E10 Myc, avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
| Mouse | |
| 48.8 kDa | |
| 0.1 mL | |
| Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| 4609 | |
| Human, Mouse, Bovine, Drosophila | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In situ PLA | |
| 90000000000 | |
| Unconjugated | |
| P01106 | |
| MYC | |
| A synthetic peptide corresponding to amino acids 408-439 (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) of human c-Myc Antibody (9E10). [UniProt# P01106] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Specific for the c-myc protein in random coil configuration, not as a helix. 9E10 does not react with V-myc. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction