missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GADD34 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 572.00 €
Specifications
| Antigen | GADD34 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18401901
|
Novus Biologicals
NBP1-89800 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18421871
|
Novus Biologicals
NBP1-89800-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GADD34 Polyclonal antibody specifically detects GADD34 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| GADD34 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Lipid and Metabolism, Unfolded Protein Response | |
| PBS (pH 7.2) and 40% Glycerol | |
| GADD34Growth arrest and DNA damage-inducible protein GADD34, growth arrest and DNA-damage-inducible 34, Myeloid differentiation primary response protein MyD116 homolog, protein phosphatase 1 regulatory subunit 15A, protein phosphatase 1, regulatory (inhibitor) subunit 15A | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: QPGEDTEEEEDEDSDTGSAEDEREAETSASTPPASAFLKAWVYRPGEDTEEEEDEDVDSEDKEDDSEAALGEAESDPHPSHPDQRAHFRGWGYRPGKETEEEEAAEDWGEAEPCP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| O75807 | |
| 23645 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title