missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis)

Product Code. 10237330 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10237330

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19598-90-4 | GdN3O9·6H2O | 451.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19598-90-4
Molecular Formula GdN3O9·6H2O
Molecular Weight (g/mol) 451.4
MDL Number MFCD00149728
InChI Key XWFVFZQEDMDSET-UHFFFAOYSA-N
Synonym gadolinium iii nitrate hexahydrate, gadolinium nitrate hexahydrate, gadolinium iii nitrate, hexahydrate 1:3:6, nitric acid, gadolinium 3+ salt, hexahydrate, gadoliniumnitratehexahydrate, gd.3no3.6h2o, gadolinium 3+ trinitrate hexahydrate, gadolinium nitrate, hexahydrate,, nitric acid,gadolinium 3+ salt, hexahydrate 8ci,9ci, gadolinium iii nitrate hexahydrate trace metals basis
PubChem CID 209258
IUPAC Name gadolinium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Gd+3]

Specifications

Melting Point 91°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 99.90%
Linear Formula Gd(NO3)3·6H2O
Solubility Information Solubility in water: soluble
Formula Weight 451.4
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gadolinium(III) nitrate hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Gadolinium(III) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.