missing translation for 'onlineSavingsMsg'
Learn More

Galactose 1-phosphate Potassium salt, MedChemExpress

Product Code. 16451037
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451037 50 mg 50mg
16444758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451037 Supplier Medchem Express Supplier No. HY113143A
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Galactose 1-phosphate Potassium salt is is an intermediate in the galactose metabolism and nucleotide sugars.

Galactose 1-phosphate Potassium salt is is an intermediate in the galactose metabolism and nucleotide sugars.

Specifications

Chemical Name or Material Galactose 1-phosphate Potassium salt
Physical Form Solid
CAS 19046-60-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C6H11K2O9P
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information H2O : 125 mg/mL (371.67 mM; Need ultrasonic)
SMILES O=P(O[K])(O[K])O[C@H]1O[C@@H]([C@H](O)[C@H](O)[C@H]1O)CO
Molecular Weight (g/mol) 336.32
Formula Weight 336.32
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.