missing translation for 'onlineSavingsMsg'
Learn More

Galangin, MedChemExpress

Product Code. 16479066
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479066 100 mg 100mg
16444827 50 mg 50mg
16413866 25 mg 25mg
16408968 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16479066 Supplier Medchem Express Supplier No. HYN0382
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Galangin (Norizalpinin) is an agonist/antagonist of the arylhydrocarbon receptor. Galangin (Norizalpinin) also shows inhibition of CYP1A1 activity.

Galangin (Norizalpinin) is an agonist/antagonist of the arylhydrocarbon receptor. Galangin (Norizalpinin) also shows inhibition of CYP1A1 activity.

Specifications

Chemical Name or Material Galangin
CAS 548-83-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O5
Quantity 100 mg
Synonym Norizalpinin 3,5,7-Trihydroxyflavone
Solubility Information DMSO : 125 mg/mL (462.55 mM; Need ultrasonic)
SMILES O=C1C(O)=C(C2=CC=CC=C2)OC3=CC(O)=CC(O)=C13
Molecular Weight (g/mol) 270.24
Formula Weight 270.24
Percent Purity 98.57%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.