missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galanin R1/GALR1 Antibody (613207) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10353B
This item is not returnable.
View return policy
Description
Galanin R1/GALR1 Monoclonal antibody specifically detects Galanin R1/GALR1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Galanin R1/GALR1 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 613207 | |
| Flow Cytometry, CyTOF-ready | |
| galanin receptor 1, galanin receptor type 1, GALNRGALNR1GAL1-R, GALR-1 | |
| NS0 mouse myeloma cell line transfected with human Galanin R1/GALR1, Accession # P47211 | |
| 0.1 mL | |
| GPCR | |
| 2587 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction