missing translation for 'onlineSavingsMsg'
Learn More

Galanthaminone, MedChemExpress

Product Code. 16443826
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443826 25 mg 25mg
16421238 5 mg 5mg
16408808 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16443826 Supplier Medchem Express Supplier No. HYI0020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Galanthaminone (Narwedin) is a competitive and reversible cholinesterase (AChE) inhibitor; is used for the treatment of mild to moderate Alzheimer's disease and various other memory impairments.

Galanthaminone (Narwedin) is a competitive and reversible cholinesterase (AChE) inhibitor

Specifications

Chemical Name or Material Galanthaminone
CAS 510-77-0
Color White
Purity Grade Notes Research
Molecular Formula C17H19NO3
Quantity 25 mg
Synonym (-)-Narwedine Narwedin
Solubility Information DMSO : 10 mg/mL (35.05 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C=C[C@@]23CCN(C)CC4=CC=C(OC)C(O[C@@]3([H])C1)=C24
Molecular Weight (g/mol) 285.34
Formula Weight 285.34
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.