Learn More
Invitrogen™ Galectin 7 Monoclonal Antibody (950723)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA560971
Product Code. 30534189
  • 479.00 € / 100µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Differential and in situ hybridization studies indicate that this lectin is specifically expressed in keratinocytes and found mainly in stratified squamous epithelium. A duplicate copy of this gene (GeneID:653499) is found adjacent to, but on the opposite strand on chromosome 19.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Galectin 7
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P47929
LGALS7, LGALS7B
E. coli-derived recombinant human Galectin-7, Ser2-Phe136.
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
950723
Unconjugated
LGALS7
GAL7; Gal-7; galectin 7; galectin 7B; galectin-7; galectin-7B; HKL-14; keratinocyte lectin 14; lectin galactoside-binding soluble 7; lectin, galactose binding, soluble 7; lectin, galactoside binding soluble 7; lectin, galactoside binding soluble 7B; lectin, galactoside-binding, soluble, 7; lectin, galactoside-binding, soluble, 7B; LGALS7; LGALS7A; LGALS7B; p53-induced gene 1 protein; p53-induced protein 1; PI7; Pig1
Mouse
Protein A/G
RUO
3963, 653499
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized