missing translation for 'onlineSavingsMsg'
Learn More

Galidesivir, MedChemExpress

Product Code. 16491739
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491739 1 mg 1mg
16420038 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491739 Supplier Medchem Express Supplier No. HY18649A
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Galidesivir (BCX4430), an adenosine analog and a direct-acting antiviral agent, disrupts viral RNA-dependent RNA polymerase (RdRp) activity. Galidesivir is active in vitro against many RNA viral pathogens, including the filoviruses and emerging infectious agents such as MERS-CoV, SARS-CoV, and SARS-CoV-2. Galidesivir inhibits some negative-sense RNA viruses with EC50s ranging from ∼3 to ∼68 μM.

Galidesivir (BCX4430), an adenosine analog and a direct-acting antiviral agent, disrupts viral RNA-dependent RNA polymerase (RdRp) activity. Galidesivir is active in vitro against many RNA viral pathogens, including the filoviruses and emerging infectious agents such as MERS-CoV, SARS-CoV, and SARS-CoV-2. Galidesivir inhibits some negative-sense RNA viruses with EC50s ranging from ∼3 to ∼68 μM.

Specifications

Chemical Name or Material Galidesivir
Physical Form Solid
CAS 249503-25-1
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C11H15N5O3
For Use With (Application) COVID-19-anti-virus
Synonym BCX4430 Immucillin-A
Solubility Information H2O : 1.53 mg/mL (5.77 mM; Need ultrasonic and warming)
SMILES O[C@H]1[C@H](C2=CNC3=C2N=CN=C3N)N[C@H](CO)[C@H]1O
Molecular Weight (g/mol) 265.27
Formula Weight 265.27
Percent Purity 99.29%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.