missing translation for 'onlineSavingsMsg'
Learn More

Gallic acid-propyl ester, Dr. Ehrenstorfer

Product Code. 30323509
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323509 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323509 Supplier Dr. Ehrenstorfer Supplier No. DREC13998300100MG
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gallic acid-propyl ester
CAS 121-79-9
Analyte or Component Names Gallic acid-propyl ester
Shipping Condition +20°C
Molecular Formula C10 H12 O5
Certifications/Compliance ISO 17034
Synonym Propyl Gallate, 3,4,5-Trihydroxybenzoic acid propyl ester, Gallic acid propyl ester, Antioxidant PG, E 310, Gallic acid n-propyl ester, Hemostyptin, Marupi Gallate, NSC 2626, Nipa 49, Nipagallin P, Nipanox S 1, PG, Progallin P, Propyl 3,4,5-trihydroxybenzoate, Propyl gallate, Tenox PG, n-Propyl 3,4,5-trihydroxybenzoate, n-Propyl gallate
InChI Formula InChI=1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3
SMILES CCCOC(=O)c1cc(O)c(O)c(O)c1
IUPAC Name propyl 3,4,5-trihydroxybenzoate
Molecular Weight (g/mol) 212.2
Quantity 100 mg
Formula Weight 212.0685 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.