Learn More
Gallium solution for ICP/MS 10mg/L HNO3 matrix, Fisher Chemical™
BIOPHARMA
Product Code. 12977353 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
50mL
Packaging:
Bottle
Product Code. 12977353
Brand: Fisher Chemical S 620 100
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Solution Contains HNO3
Concentration 10 mg/L
Quantity 100 mL