missing translation for 'onlineSavingsMsg'
Learn More

Gamithromycin, MedChemExpress

Product Code. 16485146
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485146 100 mg 100mg
16484188 10 mg 10mg
16480126 2 mg 2mg
16470537 50 mg 50mg
16466907 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16485146 Supplier Medchem Express Supplier No. HY108365

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gamithromycin is an antimicrobial agent which can inhibit the growth of MmmSC strains B237 and Tan8 with MICs of 0.00012 and 0.00006 μg/mL, respectively.

Gamithromycin is an antimicrobial agent which can inhibit the growth of MmmSC strains B237 and Tan8 with MICs of 0.00012 and 0.00006 μg/mL, respectively.

Specifications

Chemical Name or Material Gamithromycin
Physical Form Solid
CAS 145435-72-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C40H76N2O12
For Use With (Application) COVID-19-immunoregulation
Synonym ML-1709460
Solubility Information DMSO : 100 mg/mL (128.69 mM; Need ultrasonic)
SMILES O[C@H]([C@H](C[C@@H](C)O1)N(C)C)[C@]1([H])O[C@@H]2[C@H]([C@@H]([C@H](C(O[C@@H]([C@@](C)(O)[C@H](O)[C@@H](C)CN(CCC)[C@H](C)C[C@]2(O)C)CC)=O)C)O[C@@]3([H])C[C@](C)([C@@H](O)[C@H](C)O3)OC)C
Molecular Weight (g/mol) 777.04
Formula Weight 777.04
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.