Learn More
Gamma Catenin Mouse, anti-Human, CoraLite Plus 488, Clone: 1A5D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66445-20UL
Description
The Gamma Catenin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Gamma Catenin. This antibody recognizes human, rat, pig, canine antigen. The Gamma Catenin antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| Gamma Catenin | |
| 1A5D10 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Rat, Pig, Canine | |
| 155 kDa | |
| IgG1 |
| Western Blot | |
| 1000 μg/mL | |
| ARVD12, Catenin gamma, CTNNG, Desmoplakin 3, Desmoplakin III, DP3, DPIII, junction plakoglobin, JUP, PDGB, PKGB, γ-Catenin | |
| 20 μL | |
| 3728 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |