missing translation for 'onlineSavingsMsg'
Learn More

gamma-HCH, Dr. Ehrenstorfer

Product Code. 30320273
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320273 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320273 Supplier Dr. Ehrenstorfer Supplier No. DREC14073000100MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material gamma-HCH
CAS 58-89-9
Analyte or Component Names gamma-HCH
Shipping Condition +20°C
Molecular Formula C6 H6 Cl6
Certifications/Compliance ISO 17034
Synonym 1,2,3,4,5,6-Hexachlorocyclohexane, 666, Aalindan, Aficide, Agrocide, Agrocide III, Agrocide WP, Ameisenmittel Merck, Aparasin, Aphtiria, Aplidal, Arbitex, Arcotal S, BBH, BHC, BHC (insecticide), Ben-Hex, Benhexol, Bentox 10, Benzene hexachloride, Bercema-Spritz-Lindan 50, Bexol, Celanex, Chloresene, Codechine, DBH, Detmol Extract, Devoran, Dol Granule, Drilltox-Spezial Aglukon, ENT 7,796, Entomoxan, Esoderm, Fenoform forte, Forst-Nexen, Gamacid, Gamacide, Gamacide 20, Gamene, Gamma benzene hexachloride, Gamma-HCH, Gammalin, Gammalin 20, Gammaterr, Gammaxene, Gammexane, Gamoline, Geobilan, Geolin G 3, Gexane, HCC, HCCH, HCH, HGI, Heclotox, Hexa, Hexachloran, Hexachlorane, Hexachlorocyclohexane, Hexatin, Hexaverm, Hexicide, Hexyclan, Hilbeech, Hortex, Hungaria L 7, Inexit, Jacutin, Kanodane, Kokotine, Kwell, Lasochron, Lendine, Lentox, Lidenal, Lindafor, Lindane, Lindatox, Lindex, Lindosep, Lintox, Lintox 3, Linvur, Lorexane, Mglawik L, Milbol 49, Mszycol, Neo-Scabicidol, Nexen FB, Nexit, Nexit Stark, Nexol E, Nicochloran, Omnitox, Ovadziak, Owadziak, PLK, Pedraczak, Pflanzol, Prodactif, Quellada, Sang-gamma, Scabecid, Scabene, Spritz-Rapidin, Spritzlindane, Spruehpflanzol, Streunex, TAP 85, Tri-6, Verindal Ultra, Viton, (1alpha,2alpha,3beta,4alpha,5alpha,6beta)-1,2,3,4,5,6-Hexachlorocyclohexane, gamma-1,2,3,4,5,6-Hexachlorocyclohexane, gamma-666, gamma-BHC, gamma-Benzene hexachloride, gamma-Benzohexachloride, gamma-HCH, gamma-Hexachloran, gamma-Hexachlorane, gamma-Hexachlorobenzene, gamma-Hexachlorocyclohexane, gamma-Lindane
InChI Formula InChI=1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5+,6+
SMILES Cl[C@@H]1[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@H](Cl)[C@H]1Cl
IUPAC Name 1,2,3,4,5,6-hexachlorocyclohexane
Molecular Weight (g/mol) 290.83
Quantity 100 mg
Formula Weight 287.8601 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.