Learn More
gamma tubulin Mouse anti-Canine, Human, Mouse, Rat, Clone: 3F9H8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66320-1-IG-150UL
Product Code. 16858085
  • 189.00 € / 150µL
Description
This antibody can label the centrosome structure.
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
Specifications
Specifications
gamma tubulin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P23258, P83887, P83888, Q9GKK5
TUBG1
gamma tubulin Fusion Protein Ag23913
150 μL
Primary
Canine, Human, Mouse, Rat
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
3F9H8
Unconjugated
TUBG1
Gamma 1 tubulin, GCP 1, TUBG, TUBG1, TUBGCP1, Tubulin gamma 1 chain, tubulin, gamma 1, tubulin-gamma
Mouse
Protein A
RUO
103733, 252921, 403695, 7283
-20°C